Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029569.3 | NP_083845.1 | 1, 2 | 318 | ||
GenBank mRNA | AF398966.1 | - | 1, 2 | 229 | ||
AK003770.1 | - | 1, 2 | 319 | |||
BC108405.1 | - | 1, 2 | 233 | 30288598 | ||
BC120851.1 | - | 1, 2 | 284 | 40129774 | ||
BC132007.1 | - | 1, 2 | 284 | 40130284 |