Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029789.1 | NP_084065.1 | 3, 4 | 401 | ||
GenBank mRNA | AK002626.1 | - | 3, 4 | 369 | ||
AK050161.1 | - | 3, 4 | 382 | |||
AK146106.1 | - | 3, 4 | 380 | |||
AK159325.1 | - | 3, 4 | 380 | |||
AY029532.1 | - | 3, 4 | 401 | |||
BC006847.1 | - | 3, 4 | 373 | 3598473 |