Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252487.1 | NP_001239416.1 | 2 | 226 | ||
NM_001252488.1 | NP_001239417.1 | 2 | 226 | |||
NM_029772.4 | NP_084048.1 | 2 | 226 | |||
GenBank mRNA | AK014942.1 | - | 2 | 227 | ||
AY263400.1 | - | 2 | 201 | |||
BC089518.1 | - | 326 | 6774298 | |||
HM179540.1 | - | 2 | 143 | |||
HM179541.1 | - | 2 | 143 |