Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031180.2 | NP_112457.1 | 1 | 781 | ||
GenBank mRNA | AF165170.1 | - | 1 | 781 | ||
AF165171.1 | - | 1 | 785 | |||
AF178429.1 | - | 1 | 776 | |||
AK133823.1 | - | 1 | 807 | |||
BC138008.1 | - | 1 | 870 | 8861027 | ||
BC138010.1 | - | 1 | 870 | 8861029 | ||
KC810035.1 | - | 1 | 784 |