Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000372.4 | NP_000363.1 | 5 | 1914 | ||
GenBank mRNA | J03581.1 | - | 5 | 1826 | ||
M27160.1 | - | 5 | 2334 | |||
M74314.1 | - | 5 | 1878 | |||
Y00819.1 | - | 5 | 1838 |