Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 21 | 4771 | ||
GenBank mRNA | AF190449.1 | - | 21 | 4713 | ||
AK033163.1 | - | 11 | 2591 | |||
AK052798.1 | - | 1 | 103 | |||
BC138999.1 | - | 21 | 4444 | 8862021 | ||
BC145490.1 | - | 20 | 4357 | 9054053 |