Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000390.3 | NP_000381.1 | 14 | 1726 | ||
NM_001320959.1 | NP_001307888.1 | 14 | 1723 | |||
XM_017029242.1 | XP_016884731.1 | 14 | 1722 | |||
XM_017029243.1 | XP_016884732.1 | 14 | 1813 | |||
XM_017029244.1 | XP_016884733.1 | 14 | 1711 | |||
XM_017029245.1 | XP_016884734.1 | 14 | 1813 | |||
XM_017029246.1 | XP_016884735.1 | 14 | 1692 | |||
GenBank mRNA | AK291160.1 | - | 14 | 1715 | ||
AK299323.1 | - | 11 | 1431 | |||
AK312443.1 | - | 14 | 1715 | |||
M83773.1 | - | 8 | 1042 | |||
X57637.1 | - | 7 | 808 | |||
X78121.1 | - | 14 | 1723 |