Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164311.1 | NP_001157783.1 | 8 | 1302 | ||
NM_053083.3 | NP_444313.3 | 8 | 1299 | |||
GenBank mRNA | AF338440.1 | - | 7 | 1194 | ||
AK014331.1 | - | 8 | 1302 | |||
BC068307.1 | - | 8 | 1241 | 30059745 |