Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197321.1 | NP_001184250.1 | 4 | 514 | ||
NM_053202.2 | NP_444432.1 | 5 | 666 | |||
GenBank mRNA | AB221601.1 | - | 2 | 355 | ||
AB221602.1 | - | 2 | 355 | |||
AB594192.1 | - | 6 | 1071 | |||
AF339103.1 | - | 2 | 355 | |||
AF339104.1 | - | 2 | 355 | |||
BC064764.1 | - | 4 | 500 | 30430449 |