Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053194.4 | NP_444424.1 | 10 | 2363 | ||
GenBank mRNA | AF288813.1 | - | 10 | 2362 | ||
AK031466.1 | - | 10 | 2310 | |||
AK165536.1 | - | 10 | 2340 | |||
AK167746.1 | - | 10 | 2366 | |||
AK169367.1 | - | 10 | 2327 | |||
BC005633.1 | - | 2 | 2135 | |||
BC022917.1 | - | 10 | 2340 | 4460521 |