Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000606.2 | NP_000597.2 | 1 | 200 | ||
GenBank mRNA | BC113626.1 | - | 1 | 175 | 8322678 | |
BM852158.1 | - | 1 | 200 | |||
M17263.1 | - | 1 | 151 | |||
M17999.1 | - | 1 | 186 | |||
X06465.1 | - | 1 | 186 |