Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133728.3 | NP_598489.2 | 4 | 705 | ||
GenBank mRNA | AK008875.1 | - | 4 | 622 | ||
AK031295.1 | - | 4 | 663 | |||
AK034746.1 | - | 624 | ||||
AK049194.1 | - | 4 | 652 | |||
AK050045.1 | - | 4 | 644 | |||
AK076793.1 | - | 4 | 607 | |||
AK154984.1 | - | 5 | 802 | |||
AK155030.1 | - | 4 | 644 | |||
AK168103.1 | - | 4 | 645 | |||
AK170510.1 | - | 4 | 621 | |||
AK206632.1 | - | 1 | 173 | |||
BC013617.1 | - | 4 | 525 | 3979452 |