Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039803.2 | NP_001034892.1 | 8 | 1763 | ||
NM_001170639.1 | NP_001164110.1 | 7 | 1672 | |||
NM_001170640.1 | NP_001164111.1 | 6 | 1576 | |||
NM_012119.4 | NP_036251.2 | 7 | 1700 | |||
NM_178432.3 | NP_848519.1 | 7 | 1739 | |||
XM_017014561.1 | XP_016870050.1 | 8 | 1708 | |||
XM_017014562.1 | XP_016870051.1 | 9 | 1787 | |||
XM_017014563.1 | XP_016870052.1 | 7 | 1585 | |||
XM_017014564.1 | XP_016870053.1 | 8 | 1709 | |||
GenBank mRNA | AF035013.1 | - | 7 | 1605 | ||
AF113130.1 | - | 5 | 2915 | |||
AK075325.1 | - | 7 | 1492 | |||
AK226135.1 | - | 2 | 4875 | |||
AK294193.1 | - | 7 | 1660 | |||
BC002655.2 | - | 7 | 1397 | 3605918 | ||
BX647489.1 | - | 5 | 2931 |