Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134437.3 | NP_602319.1 | 4 | 439 | ||
GenBank mRNA | AF459444.1 | - | 4 | 358 | ||
AF494210.1 | - | 4 | 499 | |||
AK133233.1 | - | 6 | 602 | |||
AK139904.1 | - | 4 | 364 | |||
BC066804.1 | - | 4 | 361 | |||
BC138629.1 | - | 4 | 394 | 8861649 |