Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144856.2 | NP_659105.2 | 2 | 514 | ||
GenBank mRNA | AB069965.1 | - | 2 | 426 | ||
AK082865.1 | - | 2 | 515 | |||
AK143949.1 | - | 2 | 502 | |||
BC013474.1 | - | 2 | 491 | 4236556 | ||
BC024119.1 | - | 2 | 478 | 5096875 | ||
BC025813.1 | - | 2 | 145 | 5097639 | ||
BC026597.1 | - | 2 | 468 | 4975360 | ||
BC026598.1 | - | 2 | 466 | 4975598 |