Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130194.1 | NP_001123666.1 | 4, 5 | 570 | ||
GenBank mRNA | AY450428.1 | - | 4, 5 | 812 | ||
BC019528.1 | - | 3, 4 | 544 | 4208860 | ||
BC031186.1 | - | 3, 4 | 531 | 4989959 | ||
BC036157.1 | - | 4, 5 | 570 | 4982413 | ||
BC036163.1 | - | 4, 5 | 611 | 4984410 |