Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145471.2 | NP_663446.1 | 5 | 746 | ||
GenBank mRNA | AK036278.1 | - | 3 | 619 | ||
AK053747.1 | - | 856 | ||||
AK132158.1 | - | 3 | 1310 | |||
AK149655.1 | - | 3 | 708 | |||
AK159836.1 | - | 3 | 689 | |||
AK172871.1 | - | 3 | 1285 | |||
AY239225.1 | - | 3 | 977 | |||
AY239226.1 | - | 5 | 659 | |||
BC022126.1 | - | 5 | 779 | 5097371 | ||
BC033482.1 | - | 2 | 713 | |||
BC138217.1 | - | 3 | 920 | 8861236 |