Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145587.2 | NP_663562.1 | 4 | 4046 | ||
GenBank mRNA | AK144566.1 | - | 2 | 2189 | ||
AK189871.1 | - | 1 | 240 | |||
AK189925.1 | - | 1 | 240 | |||
AK191025.1 | - | 1 | 240 | |||
AK191105.1 | - | 1 | 240 | |||
AK192779.1 | - | 1 | 241 | |||
AK219941.1 | - | 1 | 241 | |||
BC024114.1 | - | 4 | 3787 | 5098034 | ||
BC025837.1 | - | 4 | 3883 | 5135723 | ||
BC031759.1 | - | 4 | 4046 | 4194152 |