Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 10, 11 | 1465 | ||
GenBank mRNA | AB053218.1 | - | 10, 11 | 1167 | ||
AK155336.1 | - | 10, 11 | 1466 | |||
AK178703.1 | - | 1, 2 | 71 | |||
AK183642.1 | - | 1, 2 | 71 | |||
AK205818.1 | - | 1, 2 | 71 | |||
AK211071.1 | - | 1, 2 | 71 | |||
AK216064.1 | - | 1, 2 | 71 | |||
BC026638.1 | - | 5, 6 | 573 | 4984907 | ||
BC031613.1 | - | 4, 5 | 443 | 4216720 | ||
CX205743.1 | - | 7, 8 | 725 | |||
CX206051.1 | - | 2, 3 | 224 |