Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146073.3 | NP_666185.3 | 9 | 2353 | ||
GenBank mRNA | AF542387.1 | - | 10 | 2022 | ||
AK033799.1 | - | 295 | ||||
AK046719.1 | - | 9 | 2341 | |||
AK080845.1 | - | 9 | 2357 | |||
BC021423.1 | - | 2 | 438 | |||
BC059814.1 | - | 9 | 2396 | 6813525 | ||
BC086758.1 | - | 10 | 1404 |