Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146075.2 | NP_666187.2 | 4, 5 | 970 | ||
GenBank mRNA | AK082131.1 | - | 4, 5 | 777 | ||
AK143224.1 | - | 4, 5 | 955 | |||
AK215984.1 | - | 1, 2 | 52 | |||
AY640307.1 | - | 4, 5 | 950 | |||
BC026588.1 | - | 4, 5 | 280 | 4972900 | ||
BC050010.1 | - | 2, 3 | 1182 |