Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146105.3 | NP_666217.2 | 11 | 3358 | ||
GenBank mRNA | AK036332.1 | - | 11 | 3390 | ||
AK146681.1 | - | 11 | 3448 | |||
BC034107.1 | - | 4 | 2376 | 4503249 | ||
BC054802.1 | - | 11 | 3358 | 6837803 |