Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MOK SIRNA Assays ›
Gene Symbol:
MOK
Gene Name:
MOK protein kinase
Gene Aliases:
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location:
Chr.14: 102224450 - 102305200 on Build GRCh38
UniGene ID:
Hs.104119
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
925
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MOK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272011.1
NP_001258940.1
8
886
NM_014226.2
NP_055041.1
9
976
NR_073540.1
3
324
NR_073541.1
4
371
NR_073542.1
3
324
NR_073543.1
3
287
XM_006720225.2
XP_006720288.1
9
976
XM_006720226.2
XP_006720289.1
9
976
XM_011537052.1
XP_011535354.1
9
976
XM_011537053.2
XP_011535355.1
3
1003
XM_017021549.1
XP_016877038.1
9
973
XM_017021550.1
XP_016877039.1
8
872
XM_017021551.1
XP_016877040.1
10
916
XM_017021552.1
XP_016877041.1
9
1014
XM_017021553.1
XP_016877042.1
7
811
XM_017021554.1
XP_016877043.1
7
810
XM_017021555.1
XP_016877044.1
9
973
XM_017021557.1
XP_016877046.1
8
876
XM_017021558.1
XP_016877047.1
8
839
XM_017021559.1
XP_016877048.1
8
965
GenBank mRNA
AB022694.1
-
9
970
AK094747.1
-
6
1762
AK131542.1
-
1
69
AK302349.1
-
8
871
AK312778.1
-
9
967
BC026069.1
-
8
1387
4838296
BC043179.1
-
7
1732
5287879
BC053536.1
-
8
1931
5175851
BG184866.1
-
2
167
U46191.1
-
3
264
U46192.1
-
3
314
U46193.1
-
4
371
U46194.1
-
3
310
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
regulation of cell cycle
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0