Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153535.2 | NP_705763.1 | 2 | 1164 | ||
GenBank mRNA | AK034229.1 | - | 2 | 391 | ||
AK038552.1 | - | 2 | 379 | |||
AK140440.1 | - | 2 | 1164 | |||
AK141607.1 | - | 2 | 1107 | |||
AK198284.1 | - | 2 | 105 | |||
AK204348.1 | - | 2 | 105 | |||
AK212095.1 | - | 2 | 105 | |||
AK220518.1 | - | 2 | 400 | |||
BC035537.1 | - | 2 | 1055 | 5369152 |