Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282993.1 | NP_001269922.1 | 14 | 4993 | ||
NM_001282994.1 | NP_001269923.1 | 15 | 4899 | |||
NM_172496.3 | NP_766084.3 | 15 | 5068 | |||
GenBank mRNA | AK028833.1 | - | 8 | 1951 | ||
AK122334.1 | - | 15 | 4899 | |||
AK147366.1 | - | 14 | 4993 | |||
AK147633.1 | - | 14 | 4993 | |||
AY308745.1 | - | 15 | 4907 | |||
BC025601.1 | - | 1 | 693 | |||
BC060061.1 | - | 4 | 2632 | |||
BC145116.1 | - | 12 | 4373 | |||
BC145118.1 | - | 15 | 4908 | 9053666 | ||
BC150706.1 | - | 15 | 4908 | 9087617 |