Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126.3 | NP_001117.2 | 1, 2 | 508 | ||
GenBank mRNA | AK025514.1 | - | 1, 2 | 255 | ||
AK303886.1 | - | 2, 3 | 220 | |||
BC012356.1 | - | 1, 2 | 293 | 4635347 | ||
BG700800.1 | - | 1, 2 | 487 |