Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030007.1 | NP_001025178.1 | 4 | 745 | ||
NM_001128.5 | NP_001119.3 | 4 | 745 | |||
GenBank mRNA | AB015317.1 | - | 4 | 450 | ||
AK128078.1 | - | 7 | 1300 | |||
AK225906.1 | - | 4 | 504 | |||
AK293233.1 | - | 4 | 395 | |||
AK294546.1 | - | 4 | 504 | |||
AK299630.1 | - | 5 | 679 | |||
AK314484.1 | - | 4 | 750 | |||
BC036283.1 | - | 4 | 710 | 4158263 | ||
BU902252.1 | - | 4 | 745 | |||
Y12226.1 | - | 4 | 459 |