Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172779.4 | NP_766367.1 | 2, 3 | 564 | ||
GenBank mRNA | AK031945.1 | - | 2, 3 | 556 | ||
AK083942.1 | - | 2, 3 | 526 | |||
AK160627.1 | - | 2, 3 | 524 | |||
BC051161.1 | - | 2, 3 | 546 | 6466142 |