Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172793.2 | NP_766381.1 | 3 | 439 | ||
GenBank mRNA | AK080918.1 | - | 3 | 422 | ||
AK084528.1 | - | 3 | 440 | |||
AY177687.1 | - | 3 | 406 | |||
BC025540.1 | - | 3 | 417 | |||
BC052556.1 | - | 2 | 352 | |||
BC150808.1 | - | 3 | 396 | 9087719 | ||
BC151155.1 | - | 3 | 396 | 9088043 |