Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172739.4 | NP_766327.3 | 7 | 8326 | ||
GenBank mRNA | AK032559.1 | - | 1 | 3079 | ||
AK136766.1 | - | 6 | 5002 | |||
AK137250.1 | - | 374 |