Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172772.2 | NP_766360.2 | 7 | 1469 | ||
GenBank mRNA | AK035930.1 | - | 7 | 1470 | ||
AK046403.1 | - | 7 | 1456 | |||
AK077470.1 | - | 4 | 401 | |||
AK212033.1 | - | 3 | 251 | |||
AK214619.1 | - | 2 | 137 | |||
AK216605.1 | - | 2 | 137 | |||
AK220158.1 | - | 7 | 1498 | |||
BC049357.1 | - | 7 | 1446 | |||
BC055816.1 | - | 7 | 1197 | |||
BC058683.1 | - | 7 | 1427 | 6834871 | ||
BC069845.1 | - | 7 | 1182 |