Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172893.3 | NP_766481.2 | 2 | 608 | ||
GenBank mRNA | AK036886.1 | - | 2 | 611 | ||
AK053205.1 | - | 2 | 507 | |||
AK081528.1 | - | 2 | 507 | |||
AK156623.1 | - | 2 | 477 | |||
AK162200.1 | - | 2 | 179 | |||
AK206233.1 | - | 1 | 50 | |||
BC120733.1 | - | 2 | 441 | 40129656 | ||
BC137645.1 | - | 2 | 441 | 8860664 |