Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172859.2 | NP_766447.2 | 12 | 1379 | ||
GenBank mRNA | AK032182.1 | - | 12 | 1379 | ||
AK032632.1 | - | 12 | 1370 | |||
AK163994.1 | - | 11 | 1244 | |||
BC141156.1 | - | 12 | 1278 | 9055768 | ||
BC145464.1 | - | 11 | 1152 | 9054026 |