Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285956.1 | NP_001272885.1 | 12 | 2233 | ||
NM_001285958.1 | NP_001272887.1 | 12 | 2129 | |||
NM_172874.3 | NP_766462.2 | 12 | 2155 | |||
GenBank mRNA | AK034987.1 | - | 11 | 2686 | ||
AK161018.1 | - | 12 | 2019 | |||
AY313606.1 | - | 12 | 2155 | |||
BC052487.1 | - | 12 | 1980 | |||
BC094340.1 | - | 12 | 2233 | 30609151 |