Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023842.2 | NP_076331.2 | 24 | 9169 | ||
GenBank mRNA | AK077574.1 | - | 1 | 1671 | ||
BC054853.1 | - | 1 | 821 |