Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175495.2 | NP_780704.1 | 1 | 1620 | ||
GenBank mRNA | AK047609.1 | - | 1 | 1620 | ||
AK081232.1 | - | 1 | 953 | |||
BC118612.1 | - | 1 | 1600 | 40107174 | ||
BC118614.1 | - | 1 | 1601 | 40107172 |
Set Membership: |
G-protein coupled receptor |