Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176831.4 | NP_789801.1 | 7 | 1688 | ||
GenBank mRNA | AK020612.1 | - | 1 | 430 | ||
AK036042.1 | - | 7 | 1671 | |||
AK039846.1 | - | 2 | 1062 | |||
AK046751.1 | - | 4 | 1740 | |||
BC089351.1 | - | 6 | 1651 | 30603115 |