Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5ar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146005.1 | NP_001139477.1 | 2 | 672 | ||
NM_176912.4 | NP_795886.2 | 2 | 765 | |||
GenBank mRNA | AK053187.1 | - | 2 | 766 | ||
AK160050.1 | - | 2 | 673 | |||
BC137941.1 | - | 2 | 654 | 8860960 | ||
BC145834.1 | - | 2 | 654 | 40131137 |
Set Membership: |
G-protein coupled receptor |