Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177296.4 | NP_796270.2 | 2 | 295 | ||
GenBank mRNA | AK045143.1 | - | 2 | 323 | ||
AK049446.1 | - | 2 | 509 | |||
AK145723.1 | - | 2 | 312 | |||
BC064646.1 | - | 2 | 295 | 6313452 | ||
BC075678.1 | - | 2 | 508 | 30619729 |