Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177216.4 | NP_796190.1 | 5, 6 | 520 | ||
GenBank mRNA | AK085272.1 | - | 5, 6 | 485 | ||
BC107239.1 | - | 5, 6 | 464 | 40052161 |