Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282175.1 | NP_001269104.1 | 14 | 2021 | ||
NM_001282176.1 | NP_001269105.1 | 14 | 2045 | |||
NM_001294.3 | NP_001285.1 | 14 | 2172 | |||
GenBank mRNA | AF037339.1 | - | 14 | 2015 | ||
AK027698.1 | - | 2 | 468 | |||
AK074935.1 | - | 14 | 2016 | |||
AK075262.1 | - | 14 | 2015 | |||
AK223553.1 | - | 14 | 2016 | |||
AK293314.1 | - | 14 | 2037 | |||
AK304480.1 | - | 14 | 2004 | |||
AK316094.1 | - | 14 | 2041 | |||
BC012359.1 | - | 14 | 2005 | 4685026 | ||
KU177987.1 | - | 14 | 2000 |