Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027626.1 | NP_081902.1 | 1, 2 | 347 | ||
GenBank mRNA | AK031477.1 | - | 1, 2 | 290 | ||
AK147606.1 | - | 1, 2 | 348 | |||
BC131913.1 | - | 1, 2 | 259 | |||
BC145641.1 | - | 1, 2 | 259 |