Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177829.3 | NP_808497.1 | 9 | 845 | ||
GenBank mRNA | AK039062.1 | - | 9 | 846 | ||
AK162840.1 | - | 9 | 809 | |||
AV380905.2 | - | 4 | 381 | |||
BC049733.1 | - | 502 | ||||
DQ437330.1 | - | 9 | 845 |