Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178029.3 | NP_821172.2 | 11, 12 | 3121 | ||
GenBank mRNA | BC049883.1 | - | 4, 5 | 509 |