Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178678.4 | NP_848793.1 | 2 | 647 | ||
GenBank mRNA | AK032485.1 | - | 2 | 664 | ||
AK035773.1 | - | 2 | 644 | |||
AK035858.1 | - | 2 | 650 | |||
AK035882.1 | - | 2 | 648 | |||
AK036180.1 | - | 2 | 667 | |||
AK036366.1 | - | 2 | 641 | |||
AK163753.1 | - | 2 | 640 | |||
AY182029.1 | - | 1 | 105 | |||
BC094910.1 | - | 2 | 134 | 30044805 | ||
BC113178.1 | - | 2 | 321 | 40060681 | ||
BC113784.1 | - | 2 | 321 | 40060682 |