Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178681.4 | NP_848796.2 | 4 | 607 | ||
GenBank mRNA | AK032509.1 | - | 4 | 766 | ||
AK038544.1 | - | 4 | 335 | |||
AK044516.1 | - | 4 | 417 | |||
AK047682.1 | - | 5 | 543 | |||
AK083222.1 | - | 5 | 618 | |||
AK122355.1 | - | 3 | 95 | |||
AK139408.1 | - | 4 | 608 | |||
AK161933.1 | - | 5 | 624 | |||
BC070461.1 | - | 4 | 439 | 30649561 |