Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030007.1 | NP_001025178.1 | 13 | 1500 | ||
NM_001128.5 | NP_001119.3 | 12 | 1491 | |||
GenBank mRNA | AB015317.1 | - | 12 | 1196 | ||
AK128078.1 | - | 15 | 2046 | |||
AK225906.1 | - | 13 | 1259 | |||
AK293233.1 | - | 13 | 1150 | |||
AK299630.1 | - | 14 | 1434 | |||
AK314484.1 | - | 12 | 1496 | |||
BC036283.1 | - | 13 | 1465 | 4158263 | ||
Y12226.1 | - | 13 | 1214 |