Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB1 SIRNA Assays ›
Gene Symbol:
APBB1
Gene Name:
amyloid beta precursor protein binding family B member 1
Gene Aliases:
FE65, MGC:9072, RIR
Chromosome Location:
Chr.11: 6395124 - 6419830 on Build GRCh38
UniGene ID:
Hs.372840
Species:
Human
siRNA ID
HSS100540
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164.4
NP_001155.1
7, 8
1401
NM_001257319.2
NP_001244248.1
6, 7
728
NM_001257320.2
NP_001244249.1
6, 7
816
NM_001257321.2
NP_001244250.1
7, 8
864
NM_001257323.2
NP_001244252.1
6, 7
728
NM_001257325.2
NP_001244254.1
6, 7
624
NM_001257326.2
NP_001244255.1
6, 7
824
NM_145689.2
NP_663722.1
7, 8
1401
NR_047512.2
7, 8
1401
XM_006718209.3
XP_006718272.2
7, 8
1451
XM_006718210.3
XP_006718273.2
7, 8
1486
XM_006718211.1
XP_006718274.1
7, 8
1657
XM_006718212.3
XP_006718275.2
7, 8
1451
XM_006718213.1
XP_006718276.1
7, 8
1642
XM_006718214.3
XP_006718277.2
7, 8
1451
XM_006718215.3
XP_006718278.3
7, 8
1759
XM_011520037.2
XP_011518339.1
6, 7
1137
XM_011520038.2
XP_011518340.1
6, 7
642
XM_011520040.2
XP_011518342.1
6, 7
639
XM_011520042.1
XP_011518344.1
6, 7
632
XM_017017639.1
XP_016873128.1
7, 8
1657
XM_017017640.1
XP_016873129.1
7, 8
1657
XM_017017641.1
XP_016873130.1
7, 8
1642
XM_017017642.1
XP_016873131.1
6, 7
1137
GenBank mRNA
AF394214.1
-
7, 8
1360
AK293452.1
-
8, 9
905
AK293550.1
-
6, 7
822
AK293554.1
-
6, 7
592
AK293613.1
-
6, 7
456
AK293643.1
-
6, 7
816
AK293711.1
-
5, 6
435
AK294495.1
-
6, 7
626
AK295241.1
-
6, 7
693
AK295425.1
-
6, 7
613
AK295837.1
-
6, 7
658
AK297542.1
-
1, 2
23
AK297544.1
-
8, 9
928
AK308782.1
-
6, 7
670
AK309000.1
-
6, 7
729
AK309434.1
-
6, 7
646
AK309445.1
-
6, 7
812
AK311091.1
-
7, 8
712
AK311101.1
-
6, 7
827
AK311141.1
-
6, 7
657
AK311149.1
-
6, 7
657
AK311413.1
-
6, 7
733
AK311725.1
-
8, 9
824
AK315965.1
-
6, 7
815
BC010854.2
-
7, 8
1355
3905603
BX538185.1
-
6, 7
638
DC306210.1
-
5, 6
435
EF103274.2
-
6, 7
672
L77864.1
-
7, 8
1360
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
signal transduction
axonogenesis
response to iron ion
positive regulation of neuron projection development
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone binding
tau protein binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0