Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004314.2 | NP_004305.2 | 2 | 120 | ||
XM_011520114.2 | XP_011518416.1 | 3 | 293 | |||
XM_017017763.1 | XP_016873252.1 | 3 | 314 | |||
GenBank mRNA | BC069102.1 | - | 2 | 120 | 7216917 | |
BC111729.1 | - | 2 | 96 | 40031715 | ||
S74683.1 | - | 2 | 120 |