Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
siRNA ID
HSS101364
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
25, 26
2264
NM_001190442.1
NP_001177371.1
26, 27
2136
NM_001284212.2
NP_001271141.1
23, 24
1912
NM_001284213.2
NP_001271142.1
22, 23
1822
NM_173060.3
NP_775083.1
24, 25
1972
NR_104285.1
12, 13
1125
XM_006714696.3
XP_006714759.1
28, 29
2278
XM_006714697.3
XP_006714760.1
27, 28
2292
XM_006714698.3
XP_006714761.1
26, 27
2256
XM_006714699.3
XP_006714762.1
27, 28
2219
XM_006714700.3
XP_006714763.1
26, 27
2238
XM_006714701.3
XP_006714764.1
26, 27
2254
XM_006714702.3
XP_006714765.1
26, 27
2130
XM_006714703.3
XP_006714766.1
25, 26
2215
XM_006714704.3
XP_006714767.1
26, 27
2163
XM_006714705.3
XP_006714768.1
25, 26
2169
XM_006714706.3
XP_006714769.1
25, 26
2090
XM_006714707.3
XP_006714770.1
25, 26
2041
XM_006714708.3
XP_006714771.1
24, 25
2001
XM_006714709.3
XP_006714772.1
24, 25
1964
XM_006714710.3
XP_006714773.1
23, 24
1919
XM_006714711.3
XP_006714774.1
23, 24
1964
XM_006714712.3
XP_006714775.1
22, 23
1911
XM_006714713.3
XP_006714776.1
12, 13
1128
XM_006714714.3
XP_006714777.1
12, 13
1125
XM_006714715.3
XP_006714778.1
13, 14
1854
XM_011543654.2
XP_011541956.1
26, 27
2214
XM_011543655.2
XP_011541957.1
26, 27
2211
XM_011543656.2
XP_011541958.1
25, 26
2158
XM_011543657.2
XP_011541959.1
25, 26
2040
XM_011543658.2
XP_011541960.1
23, 24
1893
XM_017009911.1
XP_016865400.1
26, 27
2253
XM_017009912.1
XP_016865401.1
27, 28
2264
XM_017009913.1
XP_016865402.1
27, 28
2236
XM_017009914.1
XP_016865403.1
26, 27
2235
XM_017009915.1
XP_016865404.1
27, 28
2240
XM_017009916.1
XP_016865405.1
26, 27
2251
XM_017009917.1
XP_016865406.1
26, 27
2172
XM_017009918.1
XP_016865407.1
25, 26
2199
XM_017009919.1
XP_016865408.1
25, 26
2172
XM_017009920.1
XP_016865409.1
25, 26
2082
XM_017009921.1
XP_016865410.1
25, 26
2096
XM_017009922.1
XP_016865411.1
24, 25
2133
XM_017009923.1
XP_016865412.1
24, 25
2130
XM_017009924.1
XP_016865413.1
24, 25
2112
XM_017009925.1
XP_016865414.1
24, 25
2051
XM_017009926.1
XP_016865415.1
24, 25
2048
XM_017009927.1
XP_016865416.1
25, 26
2037
XM_017009928.1
XP_016865417.1
24, 25
2036
XM_017009929.1
XP_016865418.1
24, 25
1955
XM_017009930.1
XP_016865419.1
23, 24
1925
XM_017009931.1
XP_016865420.1
23, 24
1916
XM_017009932.1
XP_016865421.1
22, 23
1979
GenBank mRNA
AB208816.1
-
26, 27
2100
AF327443.1
-
21, 22
1814
AK292289.1
-
12, 13
1128
AK292356.1
-
12, 13
1101
AK296919.1
-
26, 27
2094
AK297879.1
-
24, 25
1914
AK298541.1
-
25, 26
2041
AK299384.1
-
23, 24
1672
AK299402.1
-
25, 26
2021
AK300624.1
-
22, 23
1805
AK302206.1
-
13, 14
1128
AK314290.1
-
23, 24
1917
AK315146.1
-
23, 24
1916
AK315673.1
-
12, 13
1073
AL045308.1
-
5, 6
404
AL832174.1
-
24, 25
1957
AL832349.1
-
24, 25
1960
AY189741.1
-
14, 15
1127
BC013579.1
-
25, 26
1967
3878564
BC078149.1
-
20, 21
1526
BT009783.1
-
22, 23
1749
D16217.1
-
25, 26
2034
D50827.1
-
3, 4
282
S73329.1
-
4, 5
306
U26724.2
-
16, 17
1397
U31345.1
-
14, 15
1141
U31346.1
-
4, 5
357
U38525.1
-
7, 8
600
U58996.2
-
12, 13
1104
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0